Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTCAAGAGCCCGGGGAGCATCCCAG[G/T]AGATGCTCTGTAGGCAGCGTTAGGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605906 MIM: 606665 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LDB3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
OPN4 - opsin 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001030015.2 | 722 | Intron | NP_001025186.1 | |||
NM_033282.3 | 722 | Intron | NP_150598.1 | |||
XM_017016955.1 | 722 | Intron | XP_016872444.1 | |||
XM_017016956.1 | 722 | Intron | XP_016872445.1 | |||
XM_017016957.1 | 722 | Intron | XP_016872446.1 |