Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCCCGCCAATGCGGCCCAGCCCCC[A/G]GAGAGTCAGGCCCACAGAGCATGCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602180 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR4489 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MIR4489 - microRNA 4489 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4690 - microRNA 4690 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PCNX3 - pecanex homolog 3 (Drosophila) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SIPA1 - signal-induced proliferation-associated 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006747.3 | 187 | UTR 5 | NP_006738.3 | |||
NM_153253.29 | 187 | UTR 5 | NP_694985.29 | |||
XM_005274189.2 | 187 | UTR 5 | XP_005274246.1 | |||
XM_011545214.1 | 187 | UTR 5 | XP_011543516.1 |