Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTGGCTGCCGCACCCTCCAGGGC[C/T]GTCACCACAATGCTGAGCTGTCTCT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602640 MIM: 605964 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL2-SNX15 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
ARL2-SNX15 - ARL2-SNX15 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NAALADL1 - N-acetylated alpha-linked acidic dipeptidase-like 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005468.2 | 2191 | Silent Mutation | ACA,ACG | T,T 725 | NP_005459.2 | |
XM_011544706.2 | 2191 | Silent Mutation | ACA,ACG | T,T 722 | XP_011543008.1 | |
XM_011544707.2 | 2191 | Silent Mutation | ACA,ACG | T,T 677 | XP_011543009.1 | |
XM_011544708.2 | 2191 | Silent Mutation | ACA,ACG | T,T 672 | XP_011543010.1 | |
XM_011544709.2 | 2191 | Intron | XP_011543011.1 | |||
XM_011544710.2 | 2191 | Intron | XP_011543012.1 | |||
XM_011544711.2 | 2191 | Intron | XP_011543013.1 | |||
XM_011544712.2 | 2191 | Intron | XP_011543014.1 |
SAC3D1 - SAC3 domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_013299.3 | 2191 | Intron | NP_037431.3 |
SNX15 - sorting nexin 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |