Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGAGCTGGACTCCATCCGCAGCCA[C/T]CACTGGTGCATCCAGGGCTGCAGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601175 MIM: 605964 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARL2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ARL2 - ADP ribosylation factor like GTPase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199745.1 | 464 | Silent Mutation | CAC,CAT | H,H 123 | NP_001186674.1 | |
NM_001667.3 | 464 | Silent Mutation | CAC,CAT | H,H 150 | NP_001658.2 |
ARL2-SNX15 - ARL2-SNX15 readthrough (NMD candidate) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR6879 - microRNA 6879 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SNX15 - sorting nexin 15 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |