Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGACATTGGGCCCGCCCGTGATGC[A/G]AATGACCCTGTGGATGATCGCCATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613979 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PRPF6 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PRPF6 - pre-mRNA processing factor 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012469.3 | 288 | Silent Mutation | GCA,GCG | A,A 40 | NP_036601.2 | |
XM_006723769.2 | 288 | Silent Mutation | GCA,GCG | A,A 40 | XP_006723832.1 |
SAMD10 - sterile alpha motif domain containing 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |