Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATACATTTGGTTCACTGTGAGATTC[G/T]GGGGGTACTGACAGCGGCAAACACT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603293 MIM: 613992 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BNIP3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BNIP3 - BCL2 interacting protein 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004052.3 | 2921 | Intron | NP_004043.3 |
PPP2R2D - protein phosphatase 2 regulatory subunit Bdelta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291310.1 | 2921 | Intron | NP_001278239.1 | |||
NM_018461.4 | 2921 | Intron | NP_060931.2 | |||
XM_006717914.3 | 2921 | UTR 3 | XP_006717977.1 | |||
XM_006717915.2 | 2921 | Intron | XP_006717978.1 | |||
XM_017016406.1 | 2921 | Intron | XP_016871895.1 | |||
XM_017016407.1 | 2921 | Intron | XP_016871896.1 | |||
XM_017016408.1 | 2921 | Intron | XP_016871897.1 |