Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGAGAAGTTTTTAAGCCTCGTCCTC[A/G]CCCTCCTCCTCCTCGAACTCCCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603156 MIM: 615101 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
BPHL PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
BPHL - biphenyl hydrolase-like (serine hydrolase) | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001302777.1 | 1409 | Intron | NP_001289706.1 | |||
NM_004332.3 | 1409 | Intron | NP_004323.2 |
TUBB2A - tubulin beta 2A class IIa | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001069.2 | 1409 | Silent Mutation | GGC,GGT | G,G 441 | NP_001060.1 | |
NM_001310315.1 | 1409 | Silent Mutation | GGC,GGT | G,G 356 | NP_001297244.1 |