Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616054 MIM: 608731 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ELP2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
ELP2 - elongator acetyltransferase complex subunit 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001242875.2 | 141 | Silent Mutation | GGC,GGG | G,G 27 | NP_001229804.1 | |
NM_001242876.2 | 141 | Silent Mutation | GGC,GGG | G,G 27 | NP_001229805.1 | |
NM_001242877.2 | 141 | Silent Mutation | GGC,GGG | G,G 27 | NP_001229806.1 | |
NM_001242878.2 | 141 | Silent Mutation | GGC,GGG | G,G 27 | NP_001229807.1 | |
NM_001242879.2 | 141 | Silent Mutation | GGC,GGG | G,G 27 | NP_001229808.1 | |
NM_001324465.1 | 141 | Silent Mutation | GGC,GGG | G,G 27 | NP_001311394.1 | |
NM_001324466.1 | 141 | Silent Mutation | GGC,GGG | G,G 27 | NP_001311395.1 | |
NM_001324467.1 | 141 | Silent Mutation | GGC,GGG | G,G 27 | NP_001311396.1 | |
NM_001324468.1 | 141 | UTR 5 | NP_001311397.1 | |||
NM_018255.3 | 141 | Silent Mutation | GGC,GGG | G,G 27 | NP_060725.1 |
SLC39A6 - solute carrier family 39 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099406.1 | 141 | Intron | NP_001092876.1 | |||
NM_012319.3 | 141 | Intron | NP_036451.3 | |||
XM_011525900.1 | 141 | Intron | XP_011524202.1 | |||
XM_011525901.1 | 141 | Intron | XP_011524203.1 |
Set Membership: |
HapMap |