Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGCTGAAGGGGCTCAGCTTGGTCGA[C/T]AAGGAGAACACGGTGAGCCCGCGGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180390 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
RRM2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
RRM2 - ribonucleotide reductase regulatory subunit M2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001034.3 | 150 | Silent Mutation | GAC,GAT | D,D 29 | NP_001025.1 | |
NM_001165931.1 | 150 | Silent Mutation | GAC,GAT | D,D 89 | NP_001159403.1 |