Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCCAGATATGCCTGGCTCCTGAGAC[C/T]CCCCCCAGGGGCTGCCCTCCTGTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605910 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ANGPTL4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ANGPTL4 - angiopoietin like 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001039667.2 | Intron | NP_001034756.1 | ||||
NM_139314.2 | Intron | NP_647475.1 | ||||
XM_005272484.3 | Intron | XP_005272541.1 | ||||
XM_005272485.3 | Intron | XP_005272542.1 |
RAB11B-AS1 - RAB11B antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |