Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176872 MIM: 610716 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MAP2K1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MAP2K1 - mitogen-activated protein kinase kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002755.3 | Intron | NP_002746.1 | ||||
XM_011521783.2 | Intron | XP_011520085.1 | ||||
XM_017022411.1 | Intron | XP_016877900.1 | ||||
XM_017022412.1 | Intron | XP_016877901.1 | ||||
XM_017022413.1 | Intron | XP_016877902.1 |
TIPIN - TIMELESS interacting protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001289986.1 | Intron | NP_001276915.1 | ||||
NM_017858.2 | Intron | NP_060328.2 | ||||
XM_005254521.2 | Intron | XP_005254578.1 | ||||
XM_005254522.4 | Intron | XP_005254579.1 | ||||
XM_006720594.3 | Intron | XP_006720657.1 | ||||
XM_011521743.1 | Intron | XP_011520045.1 | ||||
XM_017022390.1 | Intron | XP_016877879.1 | ||||
XM_017022391.1 | Intron | XP_016877880.1 |
Set Membership: |
HapMap |