Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGGCCCCACCTGTCCCAGCCACCCC[C/T]TATGAAGCCTTCGACCCGGCAGCAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 174761 MIM: 606802 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POLD1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POLD1 - DAN polymerase delta 1, catalytic subunit | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SPIB - Spi-B transcription factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001243998.1 | 196 | Intron | NP_001230927.1 | |||
NM_001243999.1 | 196 | Silent Mutation | CCC,CCT | P,P 57 | NP_001230928.1 | |
NM_001244000.1 | 196 | Missense Mutation | CCT,CTT | P,L 38 | NP_001230929.1 | |
NM_003121.4 | 196 | Silent Mutation | CCC,CCT | P,P 57 | NP_003112.2 |