Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTGCGCTTCTTTCAAACACAAC[C/T]TGCCAGACGGAAAACCGGCTTTCCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
38 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600563 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTGFR PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTGFR - prostaglandin F receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000959.3 | 329 | Silent Mutation | ACC,ACT | T,T 21 | NP_000950.1 | |
NM_001039585.1 | 329 | Silent Mutation | ACC,ACT | T,T 21 | NP_001034674.1 | |
XM_017001873.1 | 329 | Silent Mutation | ACC,ACT | T,T 21 | XP_016857362.1 |