Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCACAAAACCAATATTTCATTCTAC[A/G]AAAATTACATCCAAACATGTGAAAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
44 submissions
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600563 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTGFR PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
PTGFR - prostaglandin F receptor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000959.3 | 701 | Silent Mutation | ACA,ACG | T,T 145 | NP_000950.1 | |
NM_001039585.1 | 701 | Silent Mutation | ACA,ACG | T,T 145 | NP_001034674.1 | |
XM_017001873.1 | 701 | Silent Mutation | ACA,ACG | T,T 145 | XP_016857362.1 |
Set Membership: |
HapMap |