Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTGGGGGATCCGGTTAGAACCTG[C/T]AGGGTGCTCTGAAGTCTGCAGTCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604721 MIM: 604504 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH2D3A PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
SH2D3A - SH2 domain containing 3A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_005490.2 | Intron | NP_005481.2 | ||||
XM_005259467.1 | Intron | XP_005259524.1 | ||||
XM_005259472.2 | Intron | XP_005259529.1 | ||||
XM_006722602.1 | Intron | XP_006722665.1 | ||||
XM_011527608.2 | Intron | XP_011525910.1 | ||||
XM_011527609.2 | Intron | XP_011525911.1 | ||||
XM_017026123.1 | Intron | XP_016881612.1 | ||||
XM_017026124.1 | Intron | XP_016881613.1 |
TRIP10 - thyroid hormone receptor interactor 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |