Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCGGGTCTGGAGGTAAATGTGTAAC[C/G]GACCCTGGTGCCCCCACCCGACCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605799 MIM: 614062 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
AMN PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
AMN - amnion associated transmembrane protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_030943.3 | 173 | Intron | NP_112205.2 | |||
XM_011537202.2 | 173 | UTR 5 | XP_011535504.1 | |||
XM_011537203.2 | 173 | Intron | XP_011535505.1 |
CDC42BPB - CDC42 binding protein kinase beta | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |