Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCATCGTGGGTGAACTCCACCCCAC[C/T]GTCTCCAGGCAGCTCCTCCACCCTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
21 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607112 MIM: 609111 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXO2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXO2 - F-box protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012168.5 | 954 | Missense Mutation | AGT,GGT | S,G 155 | NP_036300.2 |
FBXO44 - F-box protein 44 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |