Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCCAGGACCCCAGGAGGCACAGACC[C/T]CGAGGTCCATCCTGGCTCAACAGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
56 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608507 MIM: 608772 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MFN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MFN2 - mitofusin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIIP - migration and invasion inhibitory protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021933.3 | 541 | Missense Mutation | CCG,TCG | P,S 142 | NP_068752.2 | |
XM_005263487.3 | 541 | Missense Mutation | CCG,TCG | P,S 142 | XP_005263544.1 | |
XM_011541895.1 | 541 | Missense Mutation | CCG,TCG | P,S 142 | XP_011540197.1 | |
XM_011541896.1 | 541 | Missense Mutation | CCG,TCG | P,S 142 | XP_011540198.1 |
MIR6729 - microRNA 6729 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |