Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGCTTTAGCCCATTGTGTATTTGAT[C/G]AAAACTGACCAAATGGATTTCCATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 108729 MIM: 601720 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATP5C1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATP5C1 - ATP synthase, H+ transporting, mitochondrial F1 complex, gamma polypeptide 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001001973.2 | Intron | NP_001001973.1 | ||||
NM_001320886.1 | Intron | NP_001307815.1 | ||||
NM_005174.3 | Intron | NP_005165.1 | ||||
XM_017016290.1 | Intron | XP_016871779.1 |
KIN - Kin17 DNA and RNA binding protein | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |