Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CCTGGGACCATCATCTATAACCTAA[A/T]GTGTCTTGAGCCTTTACTAGAGCTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164012 MIM: 602327 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NFKB2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NFKB2 - nuclear factor kappa B subunit 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PSD - pleckstrin and Sec7 domain containing | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270965.1 | Intron | NP_001257894.1 | ||||
NM_001270966.1 | Intron | NP_001257895.1 | ||||
NM_002779.4 | Intron | NP_002770.3 | ||||
XM_011539968.1 | Intron | XP_011538270.1 | ||||
XM_011539969.2 | Intron | XP_011538271.1 | ||||
XM_017016433.1 | Intron | XP_016871922.1 | ||||
XM_017016434.1 | Intron | XP_016871923.1 |