Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTATGTTTGCCAAGTCTCCCCACTG[A/G]ATAGACGAAAGAAGATACTTTCTCT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
35 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614799 MIM: 609977 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
C1orf109 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
C1orf109 - chromosome 1 open reading frame 109 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001303030.1 | 695 | Silent Mutation | ATC,ATT | I,I 225 | NP_001289959.1 | |
NM_001303031.1 | 695 | Silent Mutation | ATC,ATT | I,I 226 | NP_001289960.1 | |
NM_017850.2 | 695 | Silent Mutation | ATC,ATT | I,I 163 | NP_060320.1 | |
XM_005270975.4 | 695 | Missense Mutation | CCA,TCA | P,S 227 | XP_005271032.1 | |
XM_005270979.1 | 695 | Missense Mutation | CCA,TCA | P,S 165 | XP_005271036.1 | |
XM_005270980.1 | 695 | Missense Mutation | CCA,TCA | P,S 165 | XP_005271037.1 | |
XM_005270981.4 | 695 | Missense Mutation | CCA,TCA | P,S 165 | XP_005271038.1 | |
XM_005270982.1 | 695 | Missense Mutation | CCA,TCA | P,S 165 | XP_005271039.1 | |
XM_011541637.1 | 695 | Missense Mutation | CCA,TCA | P,S 228 | XP_011539939.1 | |
XM_011541638.1 | 695 | Missense Mutation | CCA,TCA | P,S 228 | XP_011539940.1 | |
XM_011541639.1 | 695 | Missense Mutation | CCA,TCA | P,S 227 | XP_011539941.1 | |
XM_011541640.1 | 695 | Missense Mutation | CCA,TCA | P,S 165 | XP_011539942.1 | |
XM_011541641.1 | 695 | Missense Mutation | CCA,TCA | P,S 165 | XP_011539943.1 | |
XM_011541642.1 | 695 | Missense Mutation | CCA,TCA | P,S 165 | XP_011539944.1 | |
XM_011541643.1 | 695 | Missense Mutation | CCA,TCA | P,S 165 | XP_011539945.1 | |
XM_011541646.2 | 695 | Missense Mutation | CCA,TCA | P,S 228 | XP_011539948.1 | |
XM_017001544.1 | 695 | Missense Mutation | CCA,TCA | P,S 228 | XP_016857033.1 | |
XM_017001545.1 | 695 | Missense Mutation | CCA,TCA | P,S 165 | XP_016857034.1 | |
XM_017001546.1 | 695 | Silent Mutation | ATC,ATT | I,I 163 | XP_016857035.1 | |
XM_017001547.1 | 695 | Silent Mutation | ATC,ATT | I,I 163 | XP_016857036.1 | |
XM_017001548.1 | 695 | Silent Mutation | ATC,ATT | I,I 225 | XP_016857037.1 | |
XM_017001549.1 | 695 | Silent Mutation | ATC,ATT | I,I 225 | XP_016857038.1 | |
XM_017001550.1 | 695 | Silent Mutation | ATC,ATT | I,I 226 | XP_016857039.1 | |
XM_017001551.1 | 695 | Silent Mutation | ATC,ATT | I,I 163 | XP_016857040.1 | |
XM_017001552.1 | 695 | Silent Mutation | ATC,ATT | I,I 163 | XP_016857041.1 | |
XM_017001553.1 | 695 | Silent Mutation | ATC,ATT | I,I 163 | XP_016857042.1 | |
XM_017001554.1 | 695 | Silent Mutation | ATC,ATT | I,I 163 | XP_016857043.1 | |
XM_017001555.1 | 695 | Silent Mutation | ATC,ATT | I,I 163 | XP_016857044.1 | |
XM_017001556.1 | 695 | Silent Mutation | ATC,ATT | I,I 163 | XP_016857045.1 |
CDCA8 - cell division cycle associated 8 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |