Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAAATAAGGTTAAAAACACATATTC[C/T]TAAAAGACAAAAAAATGACACCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 146640 MIM: 601720 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITIH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITIH2 - inter-alpha-trypsin inhibitor heavy chain 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KIN - Kin17 DNA and RNA binding protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012311.3 | Intron | NP_036443.1 | ||||
XM_006717434.3 | Intron | XP_006717497.1 | ||||
XM_006717435.3 | Intron | XP_006717498.1 | ||||
XM_011519427.1 | Intron | XP_011517729.1 | ||||
XM_017015946.1 | Intron | XP_016871435.1 |