Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGCTCCTTTCCTACTTCTACTGGA[G/T]TCTCCCCGGGCGCCCCCTTGTGGTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
13 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605347 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PADI4 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PADI4 - peptidyl arginine deiminase 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012387.2 | Intron | NP_036519.2 | ||||
XM_011541150.1 | Intron | XP_011539452.1 | ||||
XM_011541151.1 | Intron | XP_011539453.1 | ||||
XM_011541152.1 | Intron | XP_011539454.1 | ||||
XM_011541153.1 | Intron | XP_011539455.1 | ||||
XM_011541154.2 | Intron | XP_011539456.1 | ||||
XM_011541155.1 | Intron | XP_011539457.1 | ||||
XM_011541156.1 | Intron | XP_011539458.1 | ||||
XM_011541157.1 | Intron | XP_011539459.1 |