Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTAATTAGAGACTGCCGCAGGGTG[G/T]GTCATAATTGTTTACGCTGTTTATG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 176882 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPRB PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PTPRB - protein tyrosine phosphatase, receptor type B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_006719528.1 | Intron | XP_006719591.1 | ||||
XM_006719529.3 | Intron | XP_006719592.1 | ||||
XM_011538614.1 | Intron | XP_011536916.1 | ||||
XM_017019723.1 | Intron | XP_016875212.1 | ||||
XM_017019724.1 | Intron | XP_016875213.1 |