Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTCCCGGACCGTATAGTTTCCAGG[A/G]CGTCTCGCCGCGCGCATGCGCAGAA
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 611077 MIM: 612048 | |||||||||||||||||||||||
Literature Links: |
CHCHD4 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
CHCHD4 - coiled-coil-helix-coiled-coil-helix domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001098502.1 | 62 | Intron | NP_001091972.1 | |||
NM_144636.2 | 62 | Intron | NP_653237.1 |
TMEM43 - transmembrane protein 43 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_024334.2 | 62 | UTR 5 | NP_077310.1 | |||
XM_017007176.1 | 62 | Intron | XP_016862665.1 |