Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAGCACTCTGCAGCGCGACGGGGC[C/T]GCTCGGGGCACCAGCGATGAGCGCG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614056 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PPP1R26 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PPP1R26 - protein phosphatase 1 regulatory subunit 26 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014811.3 | 1008 | Intron | NP_055626.3 | |||
XM_005263411.2 | 1008 | Intron | XP_005263468.1 | |||
XM_017015355.1 | 1008 | Silent Mutation | GCC,GCT | A,A 59 | XP_016870844.1 | |
XM_017015356.1 | 1008 | Silent Mutation | GCC,GCT | A,A 59 | XP_016870845.1 | |
XM_017015357.1 | 1008 | Silent Mutation | GCC,GCT | A,A 59 | XP_016870846.1 | |
XM_017015358.1 | 1008 | Silent Mutation | GCC,GCT | A,A 59 | XP_016870847.1 | |
XM_017015359.1 | 1008 | Silent Mutation | GCC,GCT | A,A 59 | XP_016870848.1 | |
XM_017015360.1 | 1008 | Silent Mutation | GCC,GCT | A,A 59 | XP_016870849.1 | |
XM_017015361.1 | 1008 | Silent Mutation | GCC,GCT | A,A 59 | XP_016870850.1 | |
XM_017015362.1 | 1008 | Silent Mutation | GCC,GCT | A,A 59 | XP_016870851.1 |