Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTGGTTCATCGCTGTCAGCTCTGAA[A/G]GAGGCTGTCCTCTCATCCTTACCCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605509 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL36A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IL36A - interleukin 36, alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014440.2 | 811 | Missense Mutation | AGA,GGA | R,G 134 | NP_055255.1 | |
XM_005263639.2 | 811 | Missense Mutation | AGA,GGA | R,G 134 | XP_005263696.1 | |
XM_011510965.1 | 811 | Missense Mutation | AGA,GGA | R,G 134 | XP_011509267.1 | |
XM_017003806.1 | 811 | Missense Mutation | AGA,GGA | R,G 134 | XP_016859295.1 |
Set Membership: |
HapMap |