Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 138571 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GYS2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GYS2 - glycogen synthase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_021957.3 | 2322 | Silent Mutation | CAC,CAT | H,H 689 | NP_068776.2 | |
XM_006719063.3 | 2322 | Silent Mutation | CAC,CAT | H,H 612 | XP_006719126.1 | |
XM_017019245.1 | 2322 | Intron | XP_016874734.1 |
SPX - spexin hormone | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |