Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CAGCTCGCTGATTGGCTGAGGCCTG[G/T]AAAGAAAGGGGAACAGGCTGGCTGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 615556 MIM: 603405 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ATAT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ATAT1 - alpha tubulin acetyltransferase 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C6orf136 - chromosome 6 open reading frame 136 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
DHX16 - DEAH-box helicase 16 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164239.1 | Intron | NP_001157711.1 | ||||
NM_003587.4 | Intron | NP_003578.2 | ||||
XM_011514938.2 | Intron | XP_011513240.1 | ||||
XM_011514939.2 | Intron | XP_011513241.1 | ||||
XM_011514940.2 | Intron | XP_011513242.1 | ||||
XM_011514941.2 | Intron | XP_011513243.1 |