Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GATCTAGAGGCAGAGCGGGTGTCAG[A/G]AGGACCCCGAGCCAGCCCCCCTCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602281 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
HAPLN3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
HAPLN3 - hyaluronan and proteoglycan link protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MFGE8 - milk fat globule-EGF factor 8 protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001114614.2 | Intron | NP_001108086.1 | ||||
NM_001310319.1 | Intron | NP_001297248.1 | ||||
NM_001310320.1 | Intron | NP_001297249.1 | ||||
NM_001310321.1 | Intron | NP_001297250.1 | ||||
NM_005928.3 | Intron | NP_005919.2 | ||||
XM_017022206.1 | Intron | XP_016877695.1 |