Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTGTTCTCCTCCCTCGCCCCGCCTT[C/T]ATCCTCCCCGTTCACGGAAACGACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609287 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LOC105378828 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LOC105378828 - uncharacterized LOC105378828 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SH3GLB1 - SH3 domain containing GRB2 like endophilin B1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206651.1 | 165 | UTR 5 | NP_001193580.1 | |||
NM_001206652.1 | 165 | UTR 5 | NP_001193581.1 | |||
NM_001206653.1 | 165 | UTR 5 | NP_001193582.1 | |||
NM_016009.4 | 165 | UTR 5 | NP_057093.1 | |||
XM_006710672.2 | 165 | UTR 5 | XP_006710735.1 | |||
XM_006710673.2 | 165 | UTR 5 | XP_006710736.1 | |||
XM_011541538.1 | 165 | UTR 5 | XP_011539840.1 | |||
XM_017001408.1 | 165 | UTR 5 | XP_016856897.1 | |||
XM_017001409.1 | 165 | UTR 5 | XP_016856898.1 | |||
XM_017001410.1 | 165 | UTR 5 | XP_016856899.1 |