Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TGTGTTTTTCACTTCTTTCTTCAGC[C/T]GCTCCACCTCCATCTTCAACAGGTC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606363 MIM: 139391 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ABI3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ABI3 - ABI family member 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
GNGT2 - G protein subunit gamma transducin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001198754.1 | 340 | Missense Mutation | CAG,CGG | Q,R 17 | NP_001185683.1 | |
NM_001198755.1 | 340 | Missense Mutation | CAG,CGG | Q,R 17 | NP_001185684.1 | |
NM_001198756.1 | 340 | Missense Mutation | CAG,CGG | Q,R 17 | NP_001185685.1 | |
NM_031498.2 | 340 | Missense Mutation | CAG,CGG | Q,R 17 | NP_113686.1 |