Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 607342 MIM: 180472 | ||||||||||||||||||||
Literature Links: |
CPEB1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CPEB1 - cytoplasmic polyadenylation element binding protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079533.1 | Intron | NP_001073001.1 | ||||
NM_001079534.1 | Intron | NP_001073002.1 | ||||
NM_001079535.1 | Intron | NP_001073003.1 | ||||
NM_001288819.1 | Intron | NP_001275748.1 | ||||
NM_001288820.1 | Intron | NP_001275749.1 | ||||
NM_030594.4 | Intron | NP_085097.3 |
RPS17 - ribosomal protein S17 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |