Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATCATTGAAAGCAAACAATTATAA[C/T]GAAGCCGTGTATTTGGTAAGTTAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600469 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
NCBP1 - nuclear cap binding protein subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002486.4 | 663 | Silent Mutation | AAC,AAT | N,N 122 | NP_002477.1 | |
XM_011518715.2 | 663 | Silent Mutation | AAC,AAT | N,N 122 | XP_011517017.1 | |
XM_017014735.1 | 663 | Silent Mutation | AAC,AAT | N,N 55 | XP_016870224.1 | |
XM_017014736.1 | 663 | UTR 5 | XP_016870225.1 | |||
XM_017014737.1 | 663 | UTR 5 | XP_016870226.1 | |||
XM_017014738.1 | 663 | UTR 5 | XP_016870227.1 |
TSTD2 - thiosulfate sulfurtransferase like domain containing 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |