Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCGGCAGGCGGAGGCAGCCCGAGG[A/G]GGCGGTTGCATGTGTGCCAGACGTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600469 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NCBP1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
NCBP1 - nuclear cap binding protein subunit 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002486.4 | 37 | UTR 5 | NP_002477.1 | |||
XM_011518715.2 | 37 | UTR 5 | XP_011517017.1 | |||
XM_017014735.1 | 37 | UTR 5 | XP_016870224.1 | |||
XM_017014736.1 | 37 | Intron | XP_016870225.1 | |||
XM_017014737.1 | 37 | Intron | XP_016870226.1 | |||
XM_017014738.1 | 37 | Intron | XP_016870227.1 |
TSTD2 - thiosulfate sulfurtransferase like domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_139246.4 | 37 | UTR 5 | NP_640339.4 | |||
XM_011518312.2 | 37 | Intron | XP_011516614.1 | |||
XM_017014342.1 | 37 | Intron | XP_016869831.1 |
Set Membership: |
HapMap |