Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCAAGTGAGTGACCCTCAGCAGCAC[A/G]AAGAACCAGGGAATGAACAGTATGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
35 submissions
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 601966 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DDI2 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DDI2 - DNA damage inducible 1 homolog 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_032341.4 | 661 | UTR 3 | NP_115717.3 |
RSC1A1 - regulatory solute carrier protein, family 1, member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006511.2 | 661 | Missense Mutation | AAA,GAA | K,E 169 | NP_006502.1 |