Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCTGGCATCCTGACAGAGGGTTCC[A/G]GTGATGGGTGCCTGGGTCCTGGTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 159465 MIM: 612192 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
MOG PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
MOG - myelin oligodendrocyte glycoprotein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001008228.2 | 1052 | Intron | NP_001008229.1 | |||
NM_001008229.2 | 1052 | Intron | NP_001008230.1 | |||
NM_001170418.1 | 1052 | Intron | NP_001163889.1 | |||
NM_002433.4 | 1052 | Intron | NP_002424.3 | |||
NM_206809.3 | 1052 | Intron | NP_996532.2 | |||
NM_206810.3 | 1052 | Intron | NP_996533.2 | |||
NM_206811.3 | 1052 | Intron | NP_996534.2 | |||
NM_206812.3 | 1052 | Intron | NP_996535.2 | |||
NM_206814.5 | 1052 | Intron | NP_996537.3 | |||
XM_005249131.3 | 1052 | Intron | XP_005249188.1 |
ZFP57 - ZFP57 zinc finger protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001109809.2 | 1052 | Silent Mutation | ACC,ACT | T,T 362 | NP_001103279.2 | |
XM_006715087.3 | 1052 | Silent Mutation | ACC,ACT | T,T 290 | XP_006715150.1 | |
XM_011514570.2 | 1052 | Silent Mutation | ACC,ACT | T,T 362 | XP_011512872.1 |