Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CTTGTTCATGCAGATGTAGAGGCCC[C/G]TCTCGGCTCCTCGGACTCGAACTCT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 600483 MIM: 606456 | ||||||||||||||||||||
Literature Links: |
FGF8 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
FGF8 - fibroblast growth factor 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001206389.1 | 500 | Missense Mutation | ACG,AGG | T,R 29 | NP_001193318.1 | |
NM_006119.4 | 500 | Missense Mutation | ACG,AGG | T,R 104 | NP_006110.1 | |
NM_033163.3 | 500 | Missense Mutation | ACG,AGG | T,R 133 | NP_149353.1 | |
NM_033164.3 | 500 | Missense Mutation | ACG,AGG | T,R 122 | NP_149354.1 | |
NM_033165.3 | 500 | Missense Mutation | ACG,AGG | T,R 93 | NP_149355.1 |
NPM3 - nucleophosmin/nucleoplasmin 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |