Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300383 MIM: 311040 MIM: 313440 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CFP PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CFP - complement factor properdin | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145252.1 | 874 | Missense Mutation | AGC,GGC | S,G 250 | NP_001138724.1 | |
NM_002621.2 | 874 | Missense Mutation | AGC,GGC | S,G 250 | NP_002612.1 | |
XM_017029575.1 | 874 | Missense Mutation | AGC,GGC | S,G 115 | XP_016885064.1 |
ELK1 - ELK1, ETS transcription factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYN1 - synapsin I | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |