Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGCAATCAGGGCGGGCCAAGGCGGC[A/T]GTTCTCGCTCCAGCTCGATGCTGCC
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 139397 | ||||||||||||||||||||
Literature Links: |
GUCY1B3 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
GUCY1B3 - guanylate cyclase 1 soluble subunit beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000857.3 | 42 | UTR 5 | NP_000848.1 | |||
NM_001291951.1 | 42 | UTR 5 | NP_001278880.1 | |||
NM_001291952.1 | 42 | UTR 5 | NP_001278881.1 | |||
NM_001291953.1 | 42 | UTR 5 | NP_001278882.1 | |||
NM_001291954.1 | 42 | UTR 5 | NP_001278883.1 | |||
NM_001291955.1 | 42 | UTR 5 | NP_001278884.1 |