Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCCAGATGCTGCTCACAGGCTCAGA[A/G]GATGGCTGCGTGTATGGCTGGGAGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARPC5L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARPC5L - actin related protein 2/3 complex subunit 5 like | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RPL35 - ribosomal protein L35 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
WDR38 - WD repeat domain 38 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001045476.2 | 205 | Silent Mutation | GAA,GAG | E,E 41 | NP_001038941.1 | |
NM_001276374.1 | 205 | Silent Mutation | GAA,GAG | E,E 41 | NP_001263303.1 | |
NM_001276375.1 | 205 | Silent Mutation | GAA,GAG | E,E 30 | NP_001263304.1 | |
NM_001276376.1 | 205 | Missense Mutation | AAG,AGG | K,R 10 | NP_001263305.1 | |
XM_005251987.3 | 205 | Silent Mutation | GAA,GAG | E,E 41 | XP_005252044.1 |