Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TTTAACAAACTGAACTCCATGGAAA[C/T]GTGAACTTTTAAAGAATTAGGAACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603921 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SUCLA2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
SUCLA2 - succinate-CoA ligase ADP-forming beta subunit | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003850.2 | Intron | NP_003841.1 | ||||
XM_017020806.1 | Intron | XP_016876295.1 | ||||
XM_017020807.1 | Intron | XP_016876296.1 | ||||
XM_017020808.1 | Intron | XP_016876297.1 | ||||
XM_017020809.1 | Intron | XP_016876298.1 |
Set Membership: |
HapMap |