Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGACTGTATGAGGCCTTGTGTCTA[C/T]AGAATGTTTTAACATTCACAACAGC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 607423 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
POMT1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
POMT1 - protein O-mannosyltransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001077365.1 | Intron | NP_001070833.1 | ||||
NM_001077366.1 | Intron | NP_001070834.1 | ||||
NM_001136113.1 | Intron | NP_001129585.1 | ||||
NM_001136114.1 | Intron | NP_001129586.1 | ||||
NM_007171.3 | Intron | NP_009102.3 | ||||
XM_005272156.1 | Intron | XP_005272213.1 | ||||
XM_005272158.1 | Intron | XP_005272215.1 | ||||
XM_005272159.1 | Intron | XP_005272216.1 | ||||
XM_005272162.2 | Intron | XP_005272219.1 | ||||
XM_006716932.1 | Intron | XP_006716995.1 | ||||
XM_011518140.1 | Intron | XP_011516442.1 | ||||
XM_011518141.1 | Intron | XP_011516443.1 | ||||
XM_011518142.1 | Intron | XP_011516444.1 | ||||
XM_011518143.1 | Intron | XP_011516445.1 | ||||
XM_011518145.1 | Intron | XP_011516447.1 | ||||
XM_017014203.1 | Intron | XP_016869692.1 | ||||
XM_017014204.1 | Intron | XP_016869693.1 | ||||
XM_017014205.1 | Intron | XP_016869694.1 |
PRRC2B - proline rich coiled-coil 2B | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |