Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTGGGGGCTGGGGCACACAGAGGGG[G/T]GTGCTGCTCACCAACAGTGCCTGAG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609362 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2G15 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PLA2G15 - phospholipase A2 group XV | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_012320.3 | Intron | NP_036452.1 | ||||
XM_005255886.3 | Intron | XP_005255943.1 | ||||
XM_011522979.2 | Intron | XP_011521281.1 | ||||
XM_011522980.2 | Intron | XP_011521282.2 |