Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATTCCAGCCCTGGTGCCTACTGCC[A/G]GAGAAAGCGGCACTGGGCTGTCCTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606967 MIM: 176847 MIM: 604119 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
LCAT PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
LCAT - lecithin-cholesterol acyltransferase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000229.1 | 3994 | Intron | NP_000220.1 |
PSMB10 - proteasome subunit beta 10 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC12A4 - solute carrier family 12 member 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145961.1 | 3994 | UTR 3 | NP_001139433.1 | |||
NM_001145962.1 | 3994 | UTR 3 | NP_001139434.1 | |||
NM_001145963.1 | 3994 | UTR 3 | NP_001139435.1 | |||
NM_001145964.1 | 3994 | UTR 3 | NP_001139436.1 | |||
NM_005072.4 | 3994 | UTR 3 | NP_005063.1 |