Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603926 MIM: 188300 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AFMID PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
AFMID - arylformamidase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
C17orf99 - chromosome 17 open reading frame 99 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SYNGR2 - synaptogyrin 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TK1 - thymidine kinase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003258.4 | Intron | NP_003249.3 | ||||
XM_017024992.1 | Intron | XP_016880481.1 |
Set Membership: |
HapMap |