Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAGTGCAGAGAAATCTGGAACGAAT[A/G]TCTTTGGCTACATTCCAGGAAATTA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605741 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
GPR183 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
GPR183 - G protein-coupled receptor 183 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004951.4 | 1024 | Missense Mutation | CAT,TAT | H,Y 284 | NP_004942.1 | |
XM_017020405.1 | 1024 | Missense Mutation | CAT,TAT | H,Y 284 | XP_016875894.1 |
UBAC2 - UBA domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144072.1 | 1024 | Intron | NP_001137544.1 | |||
NM_177967.3 | 1024 | Intron | NP_808882.1 | |||
XM_006719948.3 | 1024 | Intron | XP_006720011.1 | |||
XM_011521082.2 | 1024 | Intron | XP_011519384.1 | |||
XM_011521083.2 | 1024 | Intron | XP_011519385.1 | |||
XM_011521084.2 | 1024 | Intron | XP_011519386.1 | |||
XM_017020553.1 | 1024 | Intron | XP_016876042.1 |