Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATCAGAGAAAAGAAAGAAAAGCTGA[A/T]GAATTATTTAAGTTAGAAAGCATGG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604258 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
DLC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
DLC1 - DLC1 Rho GTPase activating protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001164271.1 | Intron | NP_001157743.1 | ||||
NM_001316668.1 | Intron | NP_001303597.1 | ||||
NM_006094.4 | Intron | NP_006085.2 | ||||
NM_024767.3 | Intron | NP_079043.3 | ||||
NM_182643.2 | Intron | NP_872584.2 | ||||
XM_005273374.1 | Intron | XP_005273431.1 | ||||
XM_017012951.1 | Intron | XP_016868440.1 | ||||
XM_017012952.1 | Intron | XP_016868441.1 |