Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAAAGAGATCAACCTGCTTCCCGAC[A/C]GAGTGCTGAGCACACCCTCCGTGCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605025 MIM: 602525 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ITGA3 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ITGA3 - integrin subunit alpha 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PDK2 - pyruvate dehydrogenase kinase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199898.1 | 218 | Silent Mutation | AGA,CGA | R,R 10 | NP_001186827.1 | |
NM_001199899.1 | 218 | Silent Mutation | AGA,CGA | R,R 10 | NP_001186828.1 | |
NM_001199900.1 | 218 | Silent Mutation | AGA,CGA | R,R 74 | NP_001186829.1 | |
NM_002611.4 | 218 | Silent Mutation | AGA,CGA | R,R 74 | NP_002602.2 |