Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605430 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SPAG9 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SPAG9 - sperm associated antigen 9 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001130527.2 | Intron | NP_001123999.1 | ||||
NM_001130528.2 | Intron | NP_001124000.1 | ||||
NM_001251971.1 | Intron | NP_001238900.1 | ||||
NM_003971.5 | Intron | NP_003962.3 | ||||
XM_005257768.2 | Intron | XP_005257825.1 | ||||
XM_005257771.2 | Intron | XP_005257828.1 | ||||
XM_005257774.3 | Intron | XP_005257831.1 | ||||
XM_017025283.1 | Intron | XP_016880772.1 | ||||
XM_017025284.1 | Intron | XP_016880773.1 | ||||
XM_017025285.1 | Intron | XP_016880774.1 |
Set Membership: |
HapMap |